accessibility menu, dialog, popup

UserName

CAS RN 542-88-1

IUPAC Name: chloro(chloromethoxy)methane
Synonyms: oxybis[chloromethane] bis (chloromethyl) ether
Molecular Weight (g/mol): 114.95
Molecular Formula: C2H4Cl2O
InChi Key: HRQGCQVOJVTVLU-UHFFFAOYSA-N
SMILES: ClCOCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results