CAS RN 542-50-7
CAS RN 542-50-7
IUPAC Name:
heptacosan-14-one
Synonyms:
14-heptacosanone
Molecular Weight (g/mol):
394.73
Molecular Formula:
C27H54O
InChi Key:
VCZMOZVQLARCOE-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCC(=O)CCCCCCCCCCCCC
1
–
1
of
1
results
1
14-Heptacosanone, 97%
CAS: 542-50-7 Molecular Formula: C27H54O Molecular Weight (g/mol): 394.728 MDL Number: MFCD00026598 InChI Key: VCZMOZVQLARCOE-UHFFFAOYSA-N Synonym: 14-heptacosanone,ditridecyl ketone,myristone,unii-fq5vc928nn,fq5vc928nn,acmc-209leu,14-heptacosanon,di-n-tridecyl ketone PubChem CID: 10955 IUPAC Name: heptacosan-14-one SMILES: CCCCCCCCCCCCCC(=O)CCCCCCCCCCCCC