accessibility menu, dialog, popup

UserName

CH3CH3NCO

CAS RN 54132-75-1

IUPAC Name: 1-isocyanato-3,5-dimethylbenzene
Synonyms: 1-isocyanato-3,5-dimethylbenzene
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: DZSGDHNHQAJZCO-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results