accessibility menu, dialog, popup

UserName

SOOHNClCl

CAS RN 54129-15-6

IUPAC Name: N-(3,5-dichlorophenyl)benzenesulfonamide
Synonyms: N-(3,5-dichlorophenyl)benzenesulfonamide
Molecular Weight (g/mol): 302.17
Molecular Formula: C12H9Cl2NO2S
InChi Key: HMKZVAZQKOKXRZ-UHFFFAOYSA-N
SMILES: ClC1=CC(NS(=O)(=O)C2=CC=CC=C2)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results