accessibility menu, dialog, popup

UserName

CAS RN 541-59-3

KAgONOONO

CAS RN 541-59-3

IUPAC Name: potassium silver(1+) bis(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-ide)
Synonyms: maleimide
Molecular Weight (g/mol): 339.1
Molecular Formula: C8H4AgKN2O4
InChi Key: KWLWLQRUFFGWNG-UHFFFAOYSA-L
SMILES: [K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1
Percent Purity 
  • (4)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (5)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (4)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (4)