accessibility menu, dialog, popup

UserName

CAS RN 541-41-3

CH3OClO

CAS RN 541-41-3

IUPAC Name: ethyl carbonochloridate
Synonyms: ethyl chloroformate
Molecular Weight (g/mol): 108.52
Molecular Formula: C3H5ClO2
InChi Key: RIFGWPKJUGCATF-UHFFFAOYSA-N
SMILES: CCOC(Cl)=O
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (6)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (6)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (7)
  • (3)
  • (6)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)