CAS RN 5408-07-1
IUPAC Name:
ethyl 4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate
Synonyms:
ethyl 4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol):
246.1
Molecular Formula:
C9H12BrNO2
InChi Key:
POLBZQUAKJYNIF-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(C)C(Br)=C(C)N1
1
–
1
of
1
results
1
ethyle4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate, 97%, Thermo Scientific™
CAS: 5408-07-1 Molecular Formula: C9H12BrNO2 Molecular Weight (g/mol): 246.10 MDL Number: MFCD02647051 InChI Key: POLBZQUAKJYNIF-UHFFFAOYSA-N Synonym: 1h-pyrrole-2-carboxylic acid, 4-bromo-3,5-dimethyl-, ethyl ester,4-bromo-3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester,ethyl 4-bromo-3,5-dimethylpyrrole-2-carboxylate,4-bromo-3,5-dimethyl-2-ethoxycarbonyl-1h-pyrrole,ethyl 4-bromo-3,5-dimethyl-1h-pyrrole-2-carboxylat,pyrrole-2-carboxylic acid, 4-bromo-3,5-dimethyl-, ethyl ester,1h-pyrrole-2-carboxylic acid, 4-bromo-3,5-dimethyl,ethyl ester PubChem CID: 138486 IUPAC Name: ethyl 4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate SMILES: CCOC(=O)C1=C(C(=C(N1)C)Br)C