accessibility menu, dialog, popup

UserName

CAS RN 54001-36-4

CH3SNClOBr

CAS RN 54001-36-4

IUPAC Name: 2-bromo-1-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one
Synonyms: 2-bromo-1-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]ethanone
Molecular Weight (g/mol): 330.62
Molecular Formula: C12H9BrClNOS
InChi Key: MDZFMZALZSZQSM-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC=C(Cl)C=C1)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results