CAS RN 540-72-7
Sodium thiocyanate, 98+%, ACS reagent
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 SMILES: [Na+].[S-]C#N
Sodium thiocyanate, 98%, extra pure
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium;thiocyanate SMILES: [Na+].[S-]C#N
Sodium thiocyanate, 98+%, for analysis
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium;thiocyanate SMILES: [Na+].[S-]C#N
Sodium thiocyanate, 96%
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium;thiocyanate SMILES: [Na+].[S-]C#N
Sodium thiocyanate, ACS, 98.0% min
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 SMILES: [Na+].[S-]C#N
Sodium Thiocyanate, ≥98%, (ACS Reagent Grade), MP Biomedicals
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium cyanosulfanide SMILES: [Na+].[S-]C#N
Sodium Thiocyanate, Supelco™
CAS: 540-72-7
Sodium Thiocyanate, Solstice
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium;thiocyanate SMILES: [Na+].[S-]C#N
Sodium Thiocyanate, 98 to 102% (titration), Reagent Grade, Honeywell™
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 SMILES: [Na+].[S-]C#N
Sodium Thiocyanate, ≥98.0% (on dried basis, AT), Honeywell™
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 SMILES: [Na+].[S-]C#N