accessibility menu, dialog, popup

UserName

OHBr

CAS RN 540-51-2

IUPAC Name: 2-bromoethan-1-ol
Synonyms: 2-bromoethanol
Molecular Weight (g/mol): 124.97
Molecular Formula: C2H5BrO
InChi Key: LDLCZOVUSADOIV-UHFFFAOYSA-N
SMILES: OCCBr
Molecular Weight (g/mol) 
  • (7)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (3)
  • (7)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (4)
  • (2)