accessibility menu, dialog, popup

UserName

CAS RN 540-37-4

NH2I

CAS RN 540-37-4

IUPAC Name: 4-iodoaniline
Synonyms: benzenamine, 4-iodo-
Molecular Weight (g/mol): 219.03
Molecular Formula: C6H6IN
InChi Key: VLVCDUSVTXIWGW-UHFFFAOYSA-N
SMILES: NC1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)