accessibility menu, dialog, popup

UserName

CAS RN 54-42-2

(S)(R)O(R)NHOHNOOHOI

CAS RN 54-42-2

IUPAC Name: 1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: iododeoxyuridine idoxuridine, 125i-labeled liquifilm, herplex idoxuridine, 125i labeled 5-iudr OftanIDU idoxuridine, 123I labeled 123I-labeled idoxuridine 131i-labeled idoxuridine idoxuridine, 3H-labeled
Molecular Weight (g/mol): 354.1
Molecular Formula: C9H11IN2O5
InChi Key: XQFRJNBWHJMXHO-UHFFFAOYNA-N
SMILES: OCC1OC(CC1O)N1C=C(I)C(=O)NC1=O
Molecular Weight (g/mol) 
  • (5)
  • (9)
Quantity 
  • (4)
  • (1)
  • (2)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (7)
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (9)
  • (4)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (7)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)