accessibility menu, dialog, popup

UserName

CAS RN 5399-87-1

OHOOHOHNNClNN

CAS RN 5399-87-1

IUPAC Name: 2-(6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: 6-chloropurine riboside
Molecular Weight (g/mol): 286.67
Molecular Formula: C10H11ClN4O4
InChi Key: XHRJGHCQQPETRH-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1C=NC2=C(Cl)N=CN=C12
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)