accessibility menu, dialog, popup

UserName

NH3H3CCH3(S)(S)H3C(R)(R)H3C(R)O(R)(S)CH3(S)H3C(S)CH3HOOO(S)O(S)(S)OH(S)HO(R)O(R)O(S)(S)HO(S)HO(R)HOHOOOHO

CAS RN 53956-04-0

IUPAC Name: (2S,3S,4S,5R,6S)-6-{[(3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl]oxy}-5-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3,4-dihydroxyoxane-2-carboxylic acid amine
Synonyms: ammonium glycyrrhizate
Molecular Weight (g/mol): 839.97
Molecular Formula: C42H65NO16
InChi Key: ILRKKHJEINIICQ-OOFFSTKBSA-N
SMILES: N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)