accessibility menu, dialog, popup

UserName

CAS RN 538-62-5

ONHHNNN(E/Z)

CAS RN 538-62-5

IUPAC Name: 3-(phenylamino)-1-(phenylimino)urea
Synonyms: 3-(phenylamino)-1-(phenylimino)urea
Molecular Weight (g/mol): 240.27
Molecular Formula: C13H12N4O
InChi Key: ZFWAHZCOKGWUIT-UHFFFAOYSA-N
SMILES: O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)