accessibility menu, dialog, popup

UserName

CAS RN 53778-73-7

IUPAC Name: 1-methoxybutan-2-ol
Synonyms: 1-methoxybutan-2-ol
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: CSZZMFWKAQEMPB-UHFFFAOYNA-N
SMILES: CCC(O)COC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results