CAS RN 53716-82-8
CAS RN 53716-82-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cyrene, TRC
CAS: 53716-82-8 Molecular Formula: C6H8O3 Molecular Weight (g/mol): 128.13 Synonym: (1S,5R)-6,8-Dioxabicyclo[3.2.1]octan-4-one,(5R)-6,8-Dioxabicyclo[3.2.1]octan-4-one,Cyrene,Dihydrolevoglucosenone IUPAC Name: (1S,5R)-6,8-dioxabicyclo[3.2.1]octan-4-one SMILES: O=C1CC[C@H]2CO[C@@H]1O2