accessibility menu, dialog, popup

UserName

CH3OONSCH3

CAS RN 53715-64-3

IUPAC Name: ethyl 4-methyl-2-phenyl-1,3-thiazole-5-carboxylate
Synonyms: ethyl 4-methyl-2-phenyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 247.31
Molecular Formula: C13H13NO2S
InChi Key: FYPLITQTMHJFKK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(S1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results