accessibility menu, dialog, popup

UserName

CAS RN 53566-38-4

CH3SnH3CCH3FFF

CAS RN 53566-38-4

IUPAC Name: tributyl[3-(trifluoromethyl)phenyl]stannane
Synonyms: tributyl[3-(trifluoromethyl)phenyl]stannane
Molecular Weight (g/mol): 435.16
Molecular Formula: C19H31F3Sn
InChi Key: LBIOKWLHIBBLEI-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=C1)C(F)(F)F
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results