CAS RN 535-83-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Trigonelline, TRC
CAS: 535-83-1 Molecular Formula: C7 H7 N O2 Molecular Weight (g/mol): 137.14 Synonym: Pyridinium, 3-carboxy-1-methyl-, inner salt,Pyridinium, 3-carboxy-1-methyl-, hydroxide, inner salt (8CI),Betaine nicotinate,Caffearine,Coffearin,Coffearine,Gynesine,N-Methylnicotinate,N-Methylnicotinic acid,Trigenolline,Trigonelline IUPAC Name: 1-methylpyridin-1-ium-3-carboxylate SMILES: C[n+]1cccc(c1)C(=O)[O-]