accessibility menu, dialog, popup

UserName

CAS RN 535-77-3

H3CCH3H3C

CAS RN 535-77-3

IUPAC Name: 1-methyl-3-(propan-2-yl)benzene
Synonyms: M-cymene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: XCYJPXQACVEIOS-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results