accessibility menu, dialog, popup

UserName

CAS RN 5349-17-7

HBrBrON

CAS RN 5349-17-7

IUPAC Name: hydrogen 2-bromo-1-(pyridin-4-yl)ethan-1-one bromide
Synonyms: hydrogen 2-bromo-1-(pyridin-4-yl)ethanone bromide
Molecular Weight (g/mol): 280.95
Molecular Formula: C7H7Br2NO
InChi Key: RGALBQILADNMKA-UHFFFAOYSA-N
SMILES: [H+].[Br-].BrCC(=O)C1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results