accessibility menu, dialog, popup

UserName

CAS RN 53448-09-2

IUPAC Name: (2R)-1-hydroxy-4-methylpentan-2-aminium
Synonyms: (2R)-1-hydroxy-4-methylpentan-2-aminium
Molecular Weight (g/mol): 118.2
Molecular Formula: C6H16NO
InChi Key: VPSSPAXIFBTOHY-ZCFIWIBFSA-O
SMILES: CC(C)C[C@@H]([NH3+])CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results