accessibility menu, dialog, popup

UserName

CAS RN 53448-07-0

H3CO(E)

CAS RN 53448-07-0

IUPAC Name: (2E)-undec-2-enal
Synonyms: trans-2-undecenal
Molecular Weight (g/mol): 168.28
Molecular Formula: C11H20O
InChi Key: PANBRUWVURLWGY-MDZDMXLPSA-N
SMILES: CCCCCCCC\C=C\C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results