accessibility menu, dialog, popup

UserName

CAS RN 53427-65-9

HNNOH

CAS RN 53427-65-9

IUPAC Name: 2-(1,4-diazepan-1-yl)ethan-1-ol
Synonyms: 2-(1,4-diazepan-1-yl)ethanol
Molecular Weight (g/mol): 144.22
Molecular Formula: C7H16N2O
InChi Key: IXKNVDAHLCFGCV-UHFFFAOYSA-N
SMILES: OCCN1CCCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results