accessibility menu, dialog, popup

UserName

CAS RN 5337-03-1

HOOO

CAS RN 5337-03-1

IUPAC Name: oxane-4-carboxylic acid
Synonyms: oxane-4-carboxylic acid
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: AVPKHOTUOHDTLW-UHFFFAOYSA-N
SMILES: OC(=O)C1CCOCC1
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)