accessibility menu, dialog, popup

UserName

CAS RN 5333-05-1

ClHNOS

CAS RN 5333-05-1

IUPAC Name: 7-chloro-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 7-chloro-2,4-dihydro-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 199.65
Molecular Formula: C8H6ClNOS
InChi Key: JNPOLSNJHCFNCG-UHFFFAOYSA-N
SMILES: ClC1=CC=C2NC(=O)CSC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)