missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-2H-1,4-benzothiazin-3(4H)-one, 97%, Thermo Scientific™

Product Code. 11416290
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11416290

Brand: Thermo Scientific Alfa Aesar H27157.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5333-05-1
MDL Number MFCD00127344
ChEBI CHEBI:74168

Specifications

Melting Point 211°C to 213°C
Molecular Formula C5H8O5
Quantity 5 g
Synonym ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone
InChI Key CUOKHACJLGPRHD-UHFFFAOYNA-N
SMILES C(C1C(C(C(=O)O1)O)O)O
IUPAC Name (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
Molecular Weight (g/mol) 148.11
PubChem CID 111064
Formula Weight 199.66
Percent Purity 97%
Chemical Name or Material 7-Chloro-2H-1,4-benzothiazin-3(4H)-one
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.