accessibility menu, dialog, popup

UserName

CAS RN 5332-73-0

IUPAC Name: 3-methoxypropan-1-amine
Synonyms: 3-methoxypropylamine
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: FAXDZWQIWUSWJH-UHFFFAOYSA-N
SMILES: COCCCN
Boiling Point 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)