CAS RN 5332-41-2
CAS RN 5332-41-2
IUPAC Name:
sodium 3-aminonaphthalene-2,7-disulfonic acid
Synonyms:
sodium amido-R-acid
Molecular Weight (g/mol):
326.29
Molecular Formula:
C10H9NNaO6S2
InChi Key:
SBRWTQONIUBBNF-UHFFFAOYSA-N
SMILES:
[Na+].NC1=C(C=C2C=C(C=CC2=C1)S(O)(=O)=O)S(O)(=O)=O
1
–
3
of
3
results
1
3-Amino-2,7-naphthalenedisulfonic Acid Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Amino-2,7-naphthalenedisulfonic Acid Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Amino-2,7-naphthalenedisulfonic Acid Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.