accessibility menu, dialog, popup

UserName

OHO

CAS RN 533-75-5

IUPAC Name: 2-hydroxycyclohepta-2,4,6-trien-1-one
Synonyms: tropolone purpurocatechol
Molecular Weight (g/mol): 122.12
Molecular Formula: C7H6O2
InChi Key: MDYOLVRUBBJPFM-UHFFFAOYSA-N
SMILES: OC1=CC=CC=CC1=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)