CAS RN 5329-14-6
Sulfamic acid, 99%
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.09 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Amidosulfonic acid, 98+%
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Amidosulfonic acid, 99.99% (metals basis)
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis)
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Sulfamic Acid, Certified AR for Analysis, Fisher Chemical
Sulfamic Acid, 99+%, (ACS Reagent Grade), MP Biomedicals
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Sulfamic Acid, ACS Reagent Grade, Ricca Chemical
CAS: 5329-14-6 Molecular Formula: H3NO3S InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid
Sulfamic acid, ≥99.5% (alkalimetric), Solstice
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Amidosulfuric Acid, Sigma-Aldrich™
CAS: 5329-14-6 Molecular Weight (g/mol): 97.09
Amidosulfonic Acid, TRC
CAS: 5329-14-6 Molecular Formula: H3 N O3 S Molecular Weight (g/mol): 97.09 Synonym: Sulfamic acid,Alprojet W,Amidosulfonic acid,Amidosulfuric acid,Aminesulfonic acid,Aminosulfonic acid,Aminosulfuric acid,Jumbo,NSC 1871,Scale Cleen,Steradent Active Plus,Sulfamidic acid,Sulfaminic acid,Sulphamic acid IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Amidosulfuric Acid, Supelco™
CAS: 5329-14-6 Molecular Weight (g/mol): 97.09
Amidosulfuric Acid, Supelco™
CAS: 5329-14-6 Molecular Weight (g/mol): 97.09
Sulfamic Acid, 98%, Reagent grade, Honeywell™
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Sulfamic acid, Honeywell Fluka™
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
- 1
- 2