accessibility menu, dialog, popup

UserName

CAS RN 53267-93-9

HN(S)OOOOHOCH3CH3CH3H3C

CAS RN 53267-93-9

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(4-methoxyphenyl)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(4-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 295.34
Molecular Formula: C15H21NO5
InChi Key: SLWWWZWJISHVOU-UHFFFAOYNA-N
SMILES: COC1=CC=C(CC(NC(=O)OC(C)(C)C)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)