accessibility menu, dialog, popup

UserName

NaOO

CAS RN 532-32-1

IUPAC Name: sodium benzoate
Synonyms: sodium benzoate
Molecular Weight (g/mol): 144.11
Molecular Formula: C7H5NaO2
InChi Key: WXMKPNITSTVMEF-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (13)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (6)
  • (1)
Physical Form 
  • (1)
  • (4)
  • (3)
Grade 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (4)
  • (3)
  • (1)
  • (5)
  • (13)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)