accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 531-37-3

IUPAC Name: 2-methoxyphenyl benzoate
Synonyms: guaiacol benzoate
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: IZYQCDNLUPLXOO-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1OC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results