accessibility menu, dialog, popup

UserName

H3COOH3C

CAS RN 53088-69-0

IUPAC Name: methyl 2-(3-methylphenyl)acetate
Synonyms: methyl 2-(3-methylphenyl)acetate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: AWTKFTNNPQGGLX-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC(C)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results