accessibility menu, dialog, popup

UserName

CAS RN 53064-41-8

IUPAC Name: ethyl 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate
Synonyms: ethyl 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate
Molecular Weight (g/mol): 183.21
Molecular Formula: C9H13NO3
InChi Key: JDWLDZUUTIPZHV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)ON=C1CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results