CAS RN 53001-73-3
CAS RN 53001-73-3
IUPAC Name:
3-(bromomethyl)-1,2,4,5-tetrafluorobenzene
Synonyms:
3-(bromomethyl)-1,2,4,5-tetrafluorobenzene
Molecular Weight (g/mol):
243
Molecular Formula:
C7H3BrF4
InChi Key:
QHZBSXQCGIQSID-UHFFFAOYSA-N
SMILES:
FC1=CC(F)=C(F)C(CBr)=C1F
1
–
1
of
1
results
1
2,3,5,6-Tetrafluorobenzyl bromide, 98+%
CAS: 53001-73-3 Molecular Formula: C7H3BrF4 Molecular Weight (g/mol): 242.999 MDL Number: MFCD00013549 InChI Key: QHZBSXQCGIQSID-UHFFFAOYSA-N Synonym: 2,3,5,6-tetrafluorobenzyl bromide,3-bromomethyl-1,2,4,5-tetrafluorobenzene,2,3,5,6-tetrafluorobenzylbromide,benzene, 3-bromomethyl-1,2,4,5-tetrafluoro,alpha-bromo-2,3,5,6-tetrafluorotoluene,pubchem7361,acmc-1b1z8,benzene,3-bromomethyl-1,2,4,5-tetrafluoro PubChem CID: 104377 IUPAC Name: 3-(bromomethyl)-1,2,4,5-tetrafluorobenzene SMILES: C1=C(C(=C(C(=C1F)F)CBr)F)F