accessibility menu, dialog, popup

UserName

CAS RN 530-93-8

O

CAS RN 530-93-8

IUPAC Name: 1,2,3,4-tetrahydronaphthalen-2-one
Synonyms: 2-tetralone
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: KCKZIWSINLBROE-UHFFFAOYSA-N
SMILES: O=C1CCC2=CC=CC=C2C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)