accessibility menu, dialog, popup

UserName

CH3OOCH3HOOHO

CAS RN 530-57-4

IUPAC Name: 4-hydroxy-3,5-dimethoxybenzoic acid
Synonyms: syringic acid syringlicacid cedar acid 4-OH-3,5-dimethoxybenzioc acid gallic acid 3,5-dimethyl ether syringic acid cu (+2) salt (1:1)
Molecular Weight (g/mol): 198.17
Molecular Formula: C9H10O5
InChi Key: JMSVCTWVEWCHDZ-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1O)C(O)=O
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)