accessibility menu, dialog, popup

UserName

H2C

CAS RN 530-48-3

IUPAC Name: (1-phenylethenyl)benzene
Synonyms: 1,1-diphenylethylene
Molecular Weight (g/mol): 180.25
Molecular Formula: C14H12
InChi Key: ZMYIIHDQURVDRB-UHFFFAOYSA-N
SMILES: C=C(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)