accessibility menu, dialog, popup

UserName

CAS RN 53-85-0

OH(R)O(R)(R)OH(S)OHNNClCl

CAS RN 53-85-0

IUPAC Name: (2R,3R,4S,5R)-2-(5,6-dichloro-1H-1,3-benzodiazol-1-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: (2R,3R,4S,5R)-2-(5,6-dichloro-1,3-benzodiazol-1-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 319.14
Molecular Formula: C12H12Cl2N2O4
InChi Key: XHSQDZXAVJRBMX-DDHJBXDOSA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=CC(Cl)=C(Cl)C=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)