CAS RN 5296-78-6
CAS RN 5296-78-6
IUPAC Name:
ethyl 2-isocyanato-3-methylbutanoate
Synonyms:
ethyl 2-isocyanato-3-methylbutanoate
Molecular Weight (g/mol):
171.2
Molecular Formula:
C8H13NO3
InChi Key:
LXPOIQVGVGYFJA-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(N=C=O)C(C)C
1
–
1
of
1
results
1
Ethyl 2-isocyanato-3-methylbutyrate, 97%, Thermo Scientific™
CAS: 5296-78-6 Molecular Formula: C8H13NO3 Molecular Weight (g/mol): 171.20 MDL Number: MFCD00078478 InChI Key: LXPOIQVGVGYFJA-UHFFFAOYNA-N Synonym: ethyl 2-isocyanato-3-methylbutyrate,97,ethyl 2s-2-isocyanato-3-methylbutanoate PubChem CID: 7018875 SMILES: CCOC(=O)C(N=C=O)C(C)C