accessibility menu, dialog, popup

UserName

H3COI

CAS RN 529-28-2

IUPAC Name: 1-iodo-2-methoxybenzene
Synonyms: 1-iodo-2-methoxybenzene
Molecular Weight (g/mol): 234.04
Molecular Formula: C7H7IO
InChi Key: DVQWNQBEUKXONL-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1I
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)