accessibility menu, dialog, popup

UserName

CAS RN 529-19-1

CH3N

CAS RN 529-19-1

IUPAC Name: 2-methylbenzonitrile
Synonyms: 2-methylbenzonitrile
Molecular Weight (g/mol): 117.15
Molecular Formula: C8H7N
InChi Key: NWPNXBQSRGKSJB-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results