accessibility menu, dialog, popup

UserName

CAS RN 528-48-3

HOOOOHOHOH

CAS RN 528-48-3

IUPAC Name: 2-(3,4-dihydroxyphenyl)-3,7-dihydroxy-4H-chromen-4-one
Synonyms: 3,7,3',4'-tetrahydroxyflavone 5-desoxyquercetin zante fustic junger fustik viset fisetin young fustic crystals fietin fisetholz fustel
Molecular Weight (g/mol): 286.24
Molecular Formula: C15H10O6
InChi Key: XHEFDIBZLJXQHF-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)