CAS RN 52785-06-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Platinum(IV) oxide hydrate, 99.9% (metals basis), Pt 71-75%
CAS: 52785-06-5 Molecular Formula: O2Pt Molecular Weight (g/mol): 227.08 MDL Number: MFCD00066964 InChI Key: YKIOKAURTKXMSB-UHFFFAOYSA-N IUPAC Name: dioxoplatinum SMILES: O=[Pt]=O
2
Platinum(IV) oxide hydrate, typical Pt-content 79-84%
CAS: 52785-06-5 Molecular Formula: O2Pt Molecular Weight (g/mol): 227.08 MDL Number: MFCD00066964 InChI Key: YKIOKAURTKXMSB-UHFFFAOYSA-N Synonym: Adams' catalyst IUPAC Name: dioxoplatinum SMILES: O=[Pt]=O