CAS RN 5270-74-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2-(4-Chloro-3-sulfamoylbenzoyl)benzoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-[3-(Aminosulfonyl)-4-chlorobenzoyl]-benzonic Acid, TRC
CAS: 5270-74-6 Molecular Formula: C14 H10 Cl N O5 S Molecular Weight (g/mol): 339.75 Synonym: 2-[3-(Aminosulfonyl)-4-chlorobenzoyl]benzoic acid,o-(4-Chloro-3-sulfamoylbenzoyl)benzoic acid IUPAC Name: 2-(4-chloro-3-sulfamoylbenzoyl)benzoic acid SMILES: NS(=O)(=O)c1cc(ccc1Cl)C(=O)c2ccccc2C(=O)O
3
2-[3-(Aminosulfonyl)-4-chlorobenzoyl]-benzonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.