accessibility menu, dialog, popup

UserName

CuHO(R)OH(R)OH(S)OH(R)OHOOHO(R)OH(R)OH(S)OH(R)OHOO

CAS RN 527-09-3

IUPAC Name: copper(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Synonyms: copper gluconate
Molecular Weight (g/mol): 453.84
Molecular Formula: C12H22CuO14
InChi Key: OCUCCJIRFHNWBP-IYEMJOQQSA-L
SMILES: [Cu++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)