accessibility menu, dialog, popup

UserName

CAS RN 526-95-4

KHO(R)OH(R)OH(S)OH(R)OHOO

CAS RN 526-95-4

IUPAC Name: potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Synonyms: potassium gluconate
Molecular Weight (g/mol): 234.25
Molecular Formula: C6H11KO7
InChi Key: HLCFGWHYROZGBI-JJKGCWMISA-M
SMILES: [K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)